missing translation for 'onlineSavingsMsg'
Learn More

Methyl Red TS 2, Ricca Chemical

Numéro de catalogue. R5045200500
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
R5045200500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. R5045200500 Fournisseur Ricca Chemical Company Code fournisseur R5045200500A
Il en reste null

Identifiants chimiques

CAS 1310-73-2 , 7732-18-5
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Spécifications

Packaging Natural Poly Bottle
Physical Form Liquid
Color Brown/Yellow
For Use With (Equipment) Conductivity Meters and Probes
Solubility Miscible with water
Solution Type Conductivity Standard
Type Conductivity Standard
Solvent or Matrix Water
Quantity 500 mL
Percent Purity 0.005
Grade Laboratory
Chemical Name or Material Methyl Red TS 2
Afficher plus de résultats Afficher moins de résultats