missing translation for 'onlineSavingsMsg'
Learn More

Methyl Red, 0.1% Aqueous, pH 4.2 to 6.3 Pink to Yellow, Certified, LabChem™

Supplier:  LabChem LC171202

Quantity 1 L

 View more versions of this product

Catalog No. LC171202


Description

Description

Specifications

Chemical Identifiers

845-10-3 , 7732-18-5
291.286
4465632
CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)[O-].[Na+]
C15H14N3NaO2
GNTPCYMJCJNRQB-UHFFFAOYSA-M
sodium;2-[[4-(dimethylamino)phenyl]diazenyl]benzoate

Specifications

Methyl Red
845-10-3 , 7732-18-5
C15H14N3NaO2
Soluble in water
GNTPCYMJCJNRQB-UHFFFAOYSA-M
sodium;2-[[4-(dimethylamino)phenyl]diazenyl]benzoate
4465632
Certified
Nitrogen oxides; Carbon monoxide; Carbon dioxide
1 L
0.1% Aqueous, pH 4.2 to 6.3 Pink to Yellow
99.9, 0.1
C15H14N3O2Na
Passes Test
CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)[O-].[Na+]
291.286
291.28
Poly Bottle
Orange
Liquid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Solution is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions