missing translation for 'onlineSavingsMsg'
Learn More

Methyl Red, 0.1% Aqueous, pH 4.2 to 6.3 Pink to Yellow, Certified, LabChem™

Catalog No. p-5389060
Change view
Click to view available options
Quantity:
125 mL
500 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC171207 125 mL
LC171201 500 mL
LC171202 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. LC171207 Supplier LabChem Supplier No. LC171207

Chemical Identifiers

CAS 845-10-3 , 7732-18-5
Molecular Formula C15H14N3NaO2
Molecular Weight (g/mol) 291.286
InChI Key GNTPCYMJCJNRQB-UHFFFAOYSA-M
PubChem CID 4465632
IUPAC Name sodium;2-[[4-(dimethylamino)phenyl]diazenyl]benzoate
SMILES CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)[O-].[Na+]

Specifications

CAS Max % 99.9, 0.1
Linear Formula C15H14N3O2Na
Solubility Information Soluble in water
Identification Passes Test
Formula Weight 291.28
Grade Certified
Packaging Poly Bottle
Decomposition Information Nitrogen oxides; Carbon monoxide; Carbon dioxide
Color Orange
Quantity 125 mL
Physical Form Liquid
Chemical Name or Material Methyl Red, 0.1% Aqueous, pH 4.2 to 6.3 Pink to Yellow
Show More Show Less
Hazard Statement GHS H Statement
Solution is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage Room Temperature