missing translation for 'onlineSavingsMsg'
Learn More

Methyl Pyridine-2-carboxylate 98.0+%, TCI America™

Catalog No. P042325G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P042325G 25 g
P0423250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P042325G Supplier TCI America Supplier No. P042325G

Chemical Identifiers

CAS 7-6-2459
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.14
MDL Number MFCD00038038
InChI Key NMMIHXMBOZYNET-UHFFFAOYSA-N
Synonym methyl picolinate, 2-picolinic acid methyl ester, 2-pyridinecarboxylic acid, methyl ester, methyl 2-pyridinecarboxylate, picolinic acid, methyl ester, 2-carbomethoxypyridine, pyridine-2-carboxylic acid methyl ester, 2-pyridinecarboxylic acid methyl ester, picolinic acid methyl ester, methylpicolinate
PubChem CID 75569
IUPAC Name methyl pyridine-2-carboxylate
SMILES COC(=O)C1=CC=CC=N1

Specifications

Color Yellow
Boiling Point 233°C
Quantity 25 g
Formula Weight 137.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Pyridine-2-carboxylate