missing translation for 'onlineSavingsMsg'
Learn More

Methyl Purple Indicator Solution, Ricca Chemical

Catalog No. 50404
Change view
Click to view available options
Quantity:
120 mL
500 mL
1 L
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
50404 120 mL Amber Glass Bottle
504016 500 mL Amber Glass Bottle
504032 1 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 50404 Supplier Ricca Chemical Company Supplier No. 50404

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For acid-base titrations

Specifications

Chemical Name or Material Methyl Purple Indicator Solution
CAS 7732-18-5, 845-10-3
CAS Min % 98.0
CAS Max % 100.0
Packaging Amber Glass Bottle
pH Color Changes pH 4.8 (purple) - 5.4 (green)
Color Dark Green
pH >5.4
Quantity 120 mL
Physical Form Liquid
Grade Indicator
Show More Show Less