missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Methyl Purple, 0.1% w/v aq. soln.
Transition interval: pH 4.8 (purple) to pH 5.4 (green) | CAS: 1340-02-9
$79.38 - $118.22
Specifications
| Chemical Name or Material | Methyl Purple |
|---|---|
| CAS | 1340-02-9 , 7732-18-5 |
| Assay Percent Range | 0.1% w/v aqueous solution |
| MDL Number | MFCD00070843 |
| Concentration or Composition (by Analyte or Components) | Methyl purple: 0.1%; Water: 99.9% |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only