missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Methyl Purple, 0.1% w/v aq. soln.

Transition interval: pH 4.8 (purple) to pH 5.4 (green) | CAS: 1340-02-9

Supplier:  Thermo Scientific Chemicals 042753K2

 View more versions of this product

Catalog No. AA42753K2

Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1340-02-9 , 7732-18-5
MFCD00070843
23 hPa (17mm Hg) at 20°C
Methyl Purple
0.1% w/v aqueous solution
Methyl purple: 0.1%; Water: 99.9%
Liquid

Specifications

Methyl Purple
0.1% w/v aqueous solution
Methyl purple: 0.1%; Water: 99.9%
1 L
1340-02-9 , 7732-18-5
MFCD00070843
23 hPa (17mm Hg) at 20°C
Liquid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only