missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Methyl Purple, 0.1% w/v aq. soln.
Transition interval: pH 4.8 (purple) to pH 5.4 (green) | CAS: 1340-02-9
Supplier: Thermo Scientific Chemicals 042753AP
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 0.1% w/v aqueous solution | |
| Methyl purple: 0.1%; Water: 99.9% | |
| Liquid |
Specifications
| Methyl Purple | |
| 0.1% w/v aqueous solution | |
| Methyl purple: 0.1%; Water: 99.9% | |
| 500 mL |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only