missing translation for 'onlineSavingsMsg'
Learn More

Methyl Pipecolinate Hydrochloride 98.0+%, TCI America™

Numéro de catalogue. M25915G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
M25915G 5 g
M25911G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. M25915G Fournisseur TCI America Code fournisseur M25915G
Il en reste null

Identifiants chimiques

CAS 32559-18-5
Molecular Formula C7H14ClNO2
Molecular Weight (g/mol) 179.644
MDL Number MFCD22123553
InChI Key APCHKWZTSCBBJX-UHFFFAOYSA-N
Synonym Pipecolinic Acid Methyl Ester Hydrochloride, Methyl 2-Piperidinecarboxylate Hydrochloride, 2-Piperidinecarboxylic Acid Methyl Ester Hydrochloride
PubChem CID 13246231
IUPAC Name methyl piperidine-2-carboxylate;hydrochloride
SMILES COC(=O)C1CCCCN1.Cl

Spécifications

Melting Point 197°C
Color White
Quantity 5 g
Formula Weight 179.64
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Methyl Pipecolinate Hydrochloride