missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Phosphorodichloridate 95.0+%, TCI America™
$376.54 - $1114.70
Chemical Identifiers
| CAS | 677-24-7 |
|---|---|
| Molecular Formula | CH3Cl2O2P |
| Molecular Weight (g/mol) | 148.91 |
| MDL Number | MFCD00002068 |
| InChI Key | SNVCRNWSNUUGEA-UHFFFAOYSA-N |
| Synonym | Methyl Dichlorophosphate |
| PubChem CID | 69611 |
| IUPAC Name | methyl chlorophosphonochloridate |
| SMILES | COP(Cl)(Cl)=O |
Chemical Identifiers
| 677-24-7 | |
| 148.91 | |
| SNVCRNWSNUUGEA-UHFFFAOYSA-N | |
| 69611 | |
| COP(Cl)(Cl)=O |
| CH3Cl2O2P | |
| MFCD00002068 | |
| Methyl Dichlorophosphate | |
| methyl chlorophosphonochloridate |
Specifications
| 677-24-7 | |
| 63°C | |
| MFCD00002068 | |
| 3265 | |
| SNVCRNWSNUUGEA-UHFFFAOYSA-N | |
| methyl chlorophosphonochloridate | |
| 69611 | |
| ≥95.0% (T) | |
| Methyl Phosphorodichloridate |
| Yellow | |
| CH3Cl2O2P | |
| 25 g | |
| Methyl Dichlorophosphate | |
| COP(Cl)(Cl)=O | |
| 148.91 | |
| 148.91 | |
| Liquid |
Safety and Handling
TSCA : No