missing translation for 'onlineSavingsMsg'
Learn More

Methyl Phosphorodichloridate 95.0+%, TCI America™

Supplier:  TCI America M0904100G

 View more versions of this product

Catalog No. M0904100G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Methyl Phosphorodichloridate
Yellow
CH3Cl2O2P
100 g
Methyl Dichlorophosphate
COP(Cl)(Cl)=O
148.91
148.91
Liquid
677-24-7
63°C
MFCD00002068
3265
SNVCRNWSNUUGEA-UHFFFAOYSA-N
methyl chlorophosphonochloridate
69611
≥95.0% (T)

Chemical Identifiers

677-24-7
148.91
SNVCRNWSNUUGEA-UHFFFAOYSA-N
69611
COP(Cl)(Cl)=O
CH3Cl2O2P
MFCD00002068
Methyl Dichlorophosphate
methyl chlorophosphonochloridate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions