missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Phosphorodichloridate 95.0+%, TCI America™
Supplier: TCI America M090425G
Specifications
| Methyl Phosphorodichloridate | |
| Yellow | |
| CH3Cl2O2P | |
| 25 g | |
| Methyl Dichlorophosphate | |
| COP(Cl)(Cl)=O | |
| 148.91 | |
| 148.91 | |
| Liquid |
| 677-24-7 | |
| 63°C | |
| MFCD00002068 | |
| 3265 | |
| SNVCRNWSNUUGEA-UHFFFAOYSA-N | |
| methyl chlorophosphonochloridate | |
| 69611 | |
| ≥95.0% (T) |
Chemical Identifiers
| 677-24-7 | |
| 148.91 | |
| SNVCRNWSNUUGEA-UHFFFAOYSA-N | |
| 69611 | |
| COP(Cl)(Cl)=O |
| CH3Cl2O2P | |
| MFCD00002068 | |
| Methyl Dichlorophosphate | |
| methyl chlorophosphonochloridate |
Safety and Handling
TSCA : No