accessibility menu, dialog, popup

UserName

CAS RN 677-24-7

H3COPClClO

CAS RN 677-24-7

IUPAC Name: methyl chlorophosphonochloridate
Synonyms: methyl chlorophosphonochloridate
Molecular Weight (g/mol): 148.91
Molecular Formula: CH3Cl2O2P
InChi Key: SNVCRNWSNUUGEA-UHFFFAOYSA-N
SMILES: COP(Cl)(Cl)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results