missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Phenylsulfonylacetate 95.0+%, TCI America™
Supplier: TCI America P135325G
Specifications
| Methyl Phenylsulfonylacetate | |
| Yellow | |
| MFCD00007555 | |
| methyl phenylsulfonylacetate, methyl 2-phenylsulfonyl acetate, methyl phenylsulfonyl acetate, methylphenylsulfonylacetate, benzenesulfonyl-acetic acid methyl ester, acmc-1ad0h, methyl benzenesulfonylacetate, methyl2-phenylsulfonyl acetate, methyl-phenylsulfonyl acetate | |
| COC(=O)CS(=O)(=O)C1=CC=CC=C1 | |
| 214.235 | |
| 214.24 | |
| Liquid |
| 34097-60-4 | |
| C9H10O4S | |
| 25 g | |
| NLEAIFBNKPYTGN-UHFFFAOYSA-N | |
| methyl 2-(benzenesulfonyl)acetate | |
| 555619 | |
| ≥95.0% (GC) |
Chemical Identifiers
| 34097-60-4 | |
| 214.235 | |
| NLEAIFBNKPYTGN-UHFFFAOYSA-N | |
| 555619 | |
| COC(=O)CS(=O)(=O)C1=CC=CC=C1 |
| C9H10O4S | |
| MFCD00007555 | |
| methyl phenylsulfonylacetate, methyl 2-phenylsulfonyl acetate, methyl phenylsulfonyl acetate, methylphenylsulfonylacetate, benzenesulfonyl-acetic acid methyl ester, acmc-1ad0h, methyl benzenesulfonylacetate, methyl2-phenylsulfonyl acetate, methyl-phenylsulfonyl acetate | |
| methyl 2-(benzenesulfonyl)acetate |
Safety and Handling
TSCA : No