accessibility menu, dialog, popup

UserName

H3COOSOO

CAS RN 34097-60-4

IUPAC Name: methyl 2-(benzenesulfonyl)acetate
Synonyms: methyl phenylsulfonyl acetate
Molecular Weight (g/mol): 214.24
Molecular Formula: C9H10O4S
InChi Key: NLEAIFBNKPYTGN-UHFFFAOYSA-N
SMILES: COC(=O)CS(=O)(=O)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results