missing translation for 'onlineSavingsMsg'
Learn More

Methyl Phenylsulfonylacetate 95.0+%, TCI America™

Catalog No. p-7135094
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P13535G 5 g
P135325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P13535G Supplier TCI America Supplier No. P13535G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 34097-60-4
Molecular Formula C9H10O4S
Molecular Weight (g/mol) 214.235
MDL Number MFCD00007555
InChI Key NLEAIFBNKPYTGN-UHFFFAOYSA-N
Synonym methyl phenylsulfonylacetate, methyl 2-phenylsulfonyl acetate, methyl phenylsulfonyl acetate, methylphenylsulfonylacetate, benzenesulfonyl-acetic acid methyl ester, acmc-1ad0h, methyl benzenesulfonylacetate, methyl2-phenylsulfonyl acetate, methyl-phenylsulfonyl acetate
PubChem CID 555619
IUPAC Name methyl 2-(benzenesulfonyl)acetate
SMILES COC(=O)CS(=O)(=O)C1=CC=CC=C1

Specifications

Color Yellow
Quantity 5 g
Formula Weight 214.24
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Phenylsulfonylacetate