missing translation for 'onlineSavingsMsg'
Learn More

Methyl phenyl carbonate, 97%

Catalog No. AAB2523214
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2523214 25 g
AAB2523203 1 g
AAB2523206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2523214 Supplier Thermo Scientific Chemicals Supplier No. B2523214
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 13509-27-8 | C8H8O3 | 152.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 13509-27-8
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.15
MDL Number MFCD04039782
InChI Key XTBFPVLHGVYOQH-UHFFFAOYSA-N
Synonym carbonic acid, methyl phenyl ester, phenyl methyl carbonate, phenyl methoxyformate, acmc-20akf2, carbonic acid methyl phenyl
PubChem CID 139482
IUPAC Name methyl phenyl carbonate
SMILES COC(=O)OC1=CC=CC=C1

Specifications

Boiling Point 210°C to 213°C
Refractive Index 1.521
Quantity 25 g
Formula Weight 152.15
Percent Purity 97%
Chemical Name or Material Methyl phenyl carbonate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only