missing translation for 'onlineSavingsMsg'
Learn More

Methyl Pentadecafluoroheptyl Ketone 93.0+%, TCI America™

Supplier:  TCI America P14525G

Catalog No. P14525G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Methyl Pentadecafluoroheptyl Ketone
Yellow
C9H3F15O
1224
LFMZSQGESVNRLL-UHFFFAOYSA-N
3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-pentadecafluorononan-2-one
12603809
≥93.0% (GC)
754-85-8
57°C
5 g
Methyl Perfluoroheptyl Ketone, 1H,1H,1H-Pentadecafluoro-2-nonanone, 1H,1H,1H-Perfluoro-2-nonanone, 3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-Pentadecafluoro-2-nonanone
CC(=O)C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F
412.098
412.10
Liquid

Chemical Identifiers

754-85-8
412.098
Methyl Perfluoroheptyl Ketone, 1H,1H,1H-Pentadecafluoro-2-nonanone, 1H,1H,1H-Perfluoro-2-nonanone, 3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-Pentadecafluoro-2-nonanone
3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-pentadecafluorononan-2-one
C9H3F15O
LFMZSQGESVNRLL-UHFFFAOYSA-N
12603809
CC(=O)C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions