missing translation for 'onlineSavingsMsg'
Learn More

Methyl Palmitate 99.5+%, TCI America™

Catalog No. S03115G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S03115G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S03115G Supplier TCI America Supplier No. S03115G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 112-39-0
Molecular Formula C17H34O2
Molecular Weight (g/mol) 270.457
MDL Number MFCD00008994
InChI Key FLIACVVOZYBSBS-UHFFFAOYSA-N
Synonym methyl palmitate, palmitic acid methyl ester, hexadecanoic acid, methyl ester, palmitic acid, methyl ester, uniphat a60, metholene 2216, methyl n-hexadecanoate, hexadecanoic acid methyl ester, n-hexadecanoic acid methyl ester, unii-dpy8vcm98i
PubChem CID 8181
ChEBI CHEBI:69187
IUPAC Name methyl hexadecanoate
SMILES CCCCCCCCCCCCCCCC(=O)OC

Specifications

Color White
Quantity 5 g
Formula Weight 270.46
Percent Purity ≥99.5% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Methyl Palmitate [Standard Material for GC]
EINECSNumber (2)-0798
TSCA Yes