missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Orange Indicator, 0.05% (w/v), Ricca Chemical
Indicator for Acid-Base Titrations
$62.07 - $103.13
Specifications
| Chemical Name or Material | Methyl Orange Indicator |
|---|---|
| CAS | 547-58-0 , 7732-18-5 |
| CAS Min % | 98.0, 0.05 |
| CAS Max % | 100.0, 0.05 |
| Concentration | 0.05% (w/v) |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
49804
|
N/A | 120 mL | Natural Poly Bottle |
Each for $62.07
|
|
||||
|
498016
|
N/A | 500 mL | Natural Poly Bottle |
Each for $94.33
|
|
||||
|
498032
|
N/A | 1 L | Natural Poly Bottle |
Each for $103.13
|
|
||||