missing translation for 'onlineSavingsMsg'
Learn More

Methyl Orange Indicator, 0.05% (w/v), Ricca Chemical

Catalog No. 49804
Change view
Click to view available options
Quantity:
120 mL
500 mL
1 L
Packaging:
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
49804 120 mL Natural Poly Bottle
498016 500 mL Natural Poly Bottle
498032 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 49804 Supplier Ricca Chemical Company Supplier No. 49804

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Indicator for Acid-Base Titrations

Specifications

Chemical Name or Material Methyl Orange
CAS 547-58-0, 7732-18-5
CAS Min % 98.0, 0.05
CAS Max % 100.0, 0.05
Concentration 0.05% (w/v)
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution
Color Orange
pH 3 to 4
Quantity 120 mL
Physical Form Liquid
Percent Purity 0.05% W/V
Grade Indicator
Show More Show Less