missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl oleate, technical
CAS: 67762-38-3 | C19H36O2 | 296.50 g/mol
Supplier: Thermo Scientific Chemicals 414640050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Methyl oleate | |
| -20.0°C | |
| 0.8700g/mL | |
| 180°C | |
| Plastic drum | |
| 1.4510 to 1.4580 | |
| MFCD00009578 | |
| 0.87 | |
| Solubility in water: insoluble. Other solubilities: soluble in most organic solvents | |
| CCCCCCCCC=CCCCCCCCC(=O)OC | |
| 296.50 | |
| 6.829 mPa.s (20°C) | |
| 296.48 | |
| Technical |
| 67762-38-3 | |
| Yellow to Amber | |
| 186.0°C (9.0 mmHg) | |
| Authentic | |
| C19H36O2 | |
| CH3(CH2)7CHCH(CH2)7COOCH3 | |
| 5 kg | |
| methyl oleate, oleic acid methyl ester, methyl cis-9-octadecenoate, oleic acid, methyl ester, edenor metio5, methyl z-9-octadecenoate, methyl-cis-oleate, exceparl m-ol, emery, oleic acid ester, esterol 112 | |
| QYDYPVFESGNLHU-ZHACJKMWSA-N | |
| methyl (Z)-octadec-9-enoate | |
| 5364509 | |
| CHEBI:27542 | |
| ≥55% | |
| Oily Liquid |
Chemical Identifiers
| 67762-38-3 | |
| 296.50 | |
| QYDYPVFESGNLHU-ZHACJKMWSA-N | |
| 5364509 | |
| methyl (Z)-octadec-9-enoate |
| C19H36O2 | |
| MFCD00009578 | |
| methyl oleate, oleic acid methyl ester, methyl cis-9-octadecenoate, oleic acid, methyl ester, edenor metio5, methyl z-9-octadecenoate, methyl-cis-oleate, exceparl m-ol, emery, oleic acid ester, esterol 112 | |
| CHEBI:27542 | |
| CCCCCCCCC=CCCCCCCCC(=O)OC |
Safety and Handling
EINECSNumber : 203-992-5
Recommended Storage : Refrigerator +4°C
RUO – Research Use Only