accessibility menu, dialog, popup

UserName

OOCH3H3C(?)

CAS RN 67762-38-3

IUPAC Name: methyl octadec-9-enoate
Synonyms: methyl 9-octadecenoate
Molecular Weight (g/mol): 296.5
Molecular Formula: C19H36O2
InChi Key: QYDYPVFESGNLHU-ZHACJKMWSA-N
SMILES: CCCCCCCCC=CCCCCCCCC(=O)OC
Boiling Point 
  • (3)
Grade 
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)