missing translation for 'onlineSavingsMsg'
Learn More

Methyl oleate, 96%

Catalog No. AAH3135822
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3135822 100 g
AAH3135814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3135822 Supplier Thermo Scientific Chemicals Supplier No. H3135822
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 112-62-9 | C19H36O2 | 296.495 g/mol

Methyl oleate is used as a chromatographic reference standard in biochemical research. It is also used as an intermediate for detergents, emulsifiers, wetting agents, stabilizers, textile treatments, plasticizers for duplicating inks, rubbers and waxes. It finds application as lubricants and lubricant additives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl oleate is used as a chromatographic reference standard in biochemical research. It is also used as an intermediate for detergents, emulsifiers, wetting agents, stabilizers, textile treatments, plasticizers for duplicating inks, rubbers and waxes. It finds application as lubricants and lubricant additives.

Solubility
Miscible with alcohol, ether and chloroform. Immiscible with water.

Notes
Air and light sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 112-62-9
Molecular Formula C19H36O2
Molecular Weight (g/mol) 296.495
MDL Number MFCD00009578
InChI Key QYDYPVFESGNLHU-KHPPLWFESA-N
Synonym methyl oleate, oleic acid methyl ester, methyl cis-9-octadecenoate, oleic acid, methyl ester, edenor metio5, methyl z-9-octadecenoate, methyl-cis-oleate, exceparl m-ol, emery, oleic acid ester, esterol 112
PubChem CID 5364509
ChEBI CHEBI:27542
IUPAC Name methyl (Z)-octadec-9-enoate
SMILES CCCCCCCCC=CCCCCCCCC(=O)OC

Specifications

Density 0.876
Boiling Point 168°C to 170°C (2 mmHg)
Flash Point 163°C (325°F)
Refractive Index 1.452
Linear Formula CH3(CH2)7CH=CH(CH2)7CO2CH3
Quantity 100 g
Beilstein 1727037
Merck Index 14,6828
Solubility Information Miscible with alcohol,ether and chloroform. Immiscible with water.
Formula Weight 296.5
Percent Purity 96%
Chemical Name or Material Methyl oleate
Show More Show Less
EINECSNumber 203-992-5
RTECSNumber RK0895000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only