missing translation for 'onlineSavingsMsg'
Learn More

Methyl octanoate, 99%

Catalog No. AC414620250
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC414620250 25 g Glass bottle
AC414621000 100 g Glass bottle
AC414625000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC414620250 Supplier Thermo Scientific Chemicals Supplier No. 414620250

CAS: 111-11-5 | C9H18O2 | 158.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 111-11-5
Molecular Formula C9H18O2
Molecular Weight (g/mol) 158.24
MDL Number MFCD00009551
InChI Key JGHZJRVDZXSNKQ-UHFFFAOYSA-N
Synonym methyl caprylate, caprylic acid methyl ester, methyl n-octanoate, octanoic acid, methyl ester, uniphat a20, octanoic acid methyl ester, methyl octylate, methyl caprylate natural, unii-7mo740x6ql, fema no. 2728
PubChem CID 8091
ChEBI CHEBI:87432
IUPAC Name methyl octanoate
SMILES CCCCCCCC(=O)OC

Specifications

Acidity 1mg KOH/g max.
Color Colorless
Density 0.8780g/mL
Boiling Point 79.0°C (12.0 mmHg)
Flash Point 73°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4165 to 1.4185
Linear Formula CH3(CH2)6COOCH3
Quantity 25 g
Specific Gravity 0.878
Solubility Information Solubility in water: insoluble
Formula Weight 158.24
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Methyl octanoate, 99%
Show More Show Less
EINECSNumber 203-835-0
RTECSNumber RH0778000
TSCA TSCA

RUO – Research Use Only