missing translation for 'onlineSavingsMsg'
Learn More

Méthyl o-Toluate 98,0+%, TCI America™

Numéro de catalogue. T029625ML
Modifier l'affichage
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
T029625ML 25 mL
T0296500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. T029625ML Fournisseur TCI America Code fournisseur T029625ML

Identifiants chimiques

CAS 89-71-4
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.177
MDL Number MFCD00008428
InChI Key WVWZECQNFWFVFW-UHFFFAOYSA-N
Synonym methyl o-toluate, methyl orthotoluate, o-toluic acid, methyl ester, methyl o-methylbenzoate, methyl toluate, benzoic acid, 2-methyl-, methyl ester, 2-methylbenzoic acid methyl ester, o-toluic acid methyl ester, methyl-o-methyl benzoate, 2-methyl methylbenzoate
PubChem CID 33094
IUPAC Name Méthyle 2-méthylbenzoate
SMILES CC1=CC=CC=C1C(=O)OC

Spécifications

Color Jaune
Boiling Point 213°C
Quantity 25 mL
Formula Weight 150.18
Percent Purity ≥98.0% (GC)
Physical Form Liquide
Chemical Name or Material Methyl o-Toluate
EINECSNumber (3)-1380
TSCA Yes