missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl n-Octanoate 99.0+%, TCI America™
$43.67 - $108.21
Chemical Identifiers
| CAS | 111-11-5 |
|---|---|
| Molecular Formula | C9H18O2 |
| Molecular Weight (g/mol) | 158.241 |
| MDL Number | MFCD00009551 |
| InChI Key | JGHZJRVDZXSNKQ-UHFFFAOYSA-N |
| Synonym | methyl caprylate, caprylic acid methyl ester, methyl n-octanoate, octanoic acid, methyl ester, uniphat a20, octanoic acid methyl ester, methyl octylate, methyl caprylate natural, unii-7mo740x6ql, fema no. 2728 |
| PubChem CID | 8091 |
| ChEBI | CHEBI:87432 |
| IUPAC Name | methyl octanoate |
| SMILES | CCCCCCCC(=O)OC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
O003325ML
|
N/A | 25 mL |
Each for $43.67
|
|
|||||
|
O0033500ML
|
N/A | 500 mL |
Each for $108.21
|
|
|||||
Chemical Identifiers
| C9H18O2 | |
| MFCD00009551 | |
| methyl caprylate, caprylic acid methyl ester, methyl n-octanoate, octanoic acid, methyl ester, uniphat a20, octanoic acid methyl ester, methyl octylate, methyl caprylate natural, unii-7mo740x6ql, fema no. 2728 | |
| CHEBI:87432 | |
| CCCCCCCC(=O)OC |
Specifications
| 111-11-5 | |
| 193°C | |
| MFCD00009551 | |
| JGHZJRVDZXSNKQ-UHFFFAOYSA-N | |
| methyl octanoate | |
| 8091 | |
| 158.24 | |
| Liquid |
| Colorless | |
| C9H18O2 | |
| methyl caprylate, caprylic acid methyl ester, methyl n-octanoate, octanoic acid, methyl ester, uniphat a20, octanoic acid methyl ester, methyl octylate, methyl caprylate natural, unii-7mo740x6ql, fema no. 2728 | |
| CCCCCCCC(=O)OC | |
| 158.241 | |
| CHEBI:87432 | |
| ≥99.0% (GC) | |
| Methyl n-Octanoate |
Safety and Handling
EINECSNumber : (2)-0782
RTECSNumber : RH0778000
TSCA : Yes