missing translation for 'onlineSavingsMsg'
Learn More

Methyl n-Octanoate 99.0+%, TCI America™

Catalog No. O003325ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O003325ML 25 mL
O0033500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O003325ML Supplier TCI America Supplier No. O003325ML

Chemical Identifiers

CAS 111-11-5
Molecular Formula C9H18O2
Molecular Weight (g/mol) 158.241
MDL Number MFCD00009551
InChI Key JGHZJRVDZXSNKQ-UHFFFAOYSA-N
Synonym methyl caprylate, caprylic acid methyl ester, methyl n-octanoate, octanoic acid, methyl ester, uniphat a20, octanoic acid methyl ester, methyl octylate, methyl caprylate natural, unii-7mo740x6ql, fema no. 2728
PubChem CID 8091
ChEBI CHEBI:87432
IUPAC Name methyl octanoate
SMILES CCCCCCCC(=O)OC

Specifications

Color Colorless
Boiling Point 193°C
Quantity 25 mL
Formula Weight 158.24
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl n-Octanoate
EINECSNumber (2)-0782
RTECSNumber RH0778000
TSCA Yes