missing translation for 'onlineSavingsMsg'
Learn More

Methyl n-Octanoate 99.0+%, TCI America™

Supplier:  TCI America O0033500ML

 View more versions of this product

Catalog No. O0033500ML


Only null left

Description

Description

Specifications

Specifications

Methyl n-Octanoate
Colorless
C9H18O2
500 mL
JGHZJRVDZXSNKQ-UHFFFAOYSA-N
methyl octanoate
8091
158.24
Liquid
111-11-5
193°C
MFCD00009551
methyl caprylate, caprylic acid methyl ester, methyl n-octanoate, octanoic acid, methyl ester, uniphat a20, octanoic acid methyl ester, methyl octylate, methyl caprylate natural, unii-7mo740x6ql, fema no. 2728
CCCCCCCC(=O)OC
158.241
CHEBI:87432
≥99.0% (GC)

Chemical Identifiers

111-11-5
158.241
JGHZJRVDZXSNKQ-UHFFFAOYSA-N
8091
methyl octanoate
C9H18O2
MFCD00009551
methyl caprylate, caprylic acid methyl ester, methyl n-octanoate, octanoic acid, methyl ester, uniphat a20, octanoic acid methyl ester, methyl octylate, methyl caprylate natural, unii-7mo740x6ql, fema no. 2728
CHEBI:87432
CCCCCCCC(=O)OC
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0782

RTECSNumber : RH0778000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions