missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl myristoleate, ≥98.5% (GC), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 56219-06-8 |
| Shipping Condition | Ambient |
| Physical Form | Neat |
| Linear Formula | C15H28O2 |
| MDL Number | MFCD00056299 |
| Synonym | Methyl cis-9-tetradecenoate; Myristoleic acid methyl ester |
| Type | Analytical Standard |
| Quantity | 100 mg |
| Formula Weight | 240.38 |
| Percent Purity | ≥98.5% (GC) |
| Show More |