missing translation for 'onlineSavingsMsg'
Learn More

Methyl mercaptoacetate, 95%

Catalog No. AAL0378336
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0378336 500 g
AAL0378322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0378336 Supplier Thermo Scientific Chemicals Supplier No. L0378336
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2365-48-2 | C3H6O2S | 106.14 g/mol

Methyl thioglycolate was used in the preparation of 3-carbomethoxy-4- oxotetrahydrothiopyran, 2- and 4-carbomethoxy-3-oxotetrahydrothiophene. It is used to prepare methyl thioglycolate and aminoethanethiol conjugated gold nanorods.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl thioglycolate was used in the preparation of 3-carbomethoxy-4- oxotetrahydrothiopyran, 2- and 4-carbomethoxy-3-oxotetrahydrothiophene. It is used to prepare methyl thioglycolate and aminoethanethiol conjugated gold nanorods.

Solubility
Soluble in water. (40 g/L at 20°C)

Notes
Air Sensitive. Store under dry inert gas. Incompatible with oxidizing agents and air.

Chemical Identifiers

CAS 2365-48-2
Molecular Formula C3H6O2S
Molecular Weight (g/mol) 106.14
MDL Number MFCD00004873
InChI Key MKIJJIMOAABWGF-UHFFFAOYSA-N
Synonym methyl thioglycolate, methyl mercaptoacetate, methyl 2-mercaptoacetate, methylthioglycolate, thioglycolic acid methyl ester, acetic acid, mercapto-, methyl ester, methanethiol, methoxy, usaf ek-7119, thioglykolsaeure-methylester, unii-o1608la9el
PubChem CID 16907
IUPAC Name methyl 2-sulfanylacetate
SMILES COC(=O)CS

Specifications

Melting Point -3°C
Density 1.163
Boiling Point 148°C to 149°C
Flash Point 56°C (132°F)
Odor Stench
Refractive Index 1.466
Linear Formula HSCH2COOCH3
Quantity 500 g
UN Number UN1228
Beilstein 506259
Sensitivity Air Sensitive
Solubility Information Soluble in water. (40g/L at 20°C)
Formula Weight 106.14
Percent Purity 98%
Chemical Name or Material Methyl mercaptoacetate
Show More Show Less
Hazard Category H226-H301-H315-H317-H319-H332
Hazard Statement GHS H Statement
H301-H226-H315-H319-H335
Toxic if swallowed.
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P272-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P333+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: MERCAPTANS, LIQUID, FLAMMABLE, TOXIC, N.O.S.
EINECSNumber 219-121-7
RTECSNumber AI7350000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only