missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl L-(-)-Lactate 98.0+%, TCI America™
Spécifications
Spécifications
| Color | Colorless |
| Boiling Point | 140°C |
| Quantity | 25 g |
| UN Number | 3272 |
| Formula Weight | 104.11 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Methyl L-(-)-Lactate |
Sécurité et manipulation
| EINECSNumber | (2)-1370 |
| TSCA | Yes |