missing translation for 'onlineSavingsMsg'
Learn More

Methyl isobutyrylacetate, 97+%

CAS: 42558-54-3 | C7H12O3 | 144.17 g/mol

$89.14 - $198.47

Chemical Identifiers

CAS 42558-54-3
Molecular Formula C7H12O3
Molecular Weight (g/mol) 144.17
MDL Number MFCD00040499
InChI Key HNNFDXWDCFCVDM-UHFFFAOYSA-N
Synonym methyl isobutyrylacetate, methyl 4-methyl-3-oxovalerate, 4-methyl-3-oxovaleric acid methyl ester, iso-butyryl methyl acetate, methyl 4-methyl-3-oxo-pentanoate, methylisobutyrylacetate, acmc-209jq9, ksc237g2d, methyl 4-methyl-3oxopentanoate, methyl4-methyl-3-oxopentanoate
PubChem CID 2759969
IUPAC Name methyl 4-methyl-3-oxopentanoate
SMILES CC(C)C(=O)CC(=O)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAL1621614
View Documents Promotion Details
N/A 25 g
Each for $89.14
Only null left
 
AAL1621622
View Documents Promotion Details
N/A 100 g
Each for $198.47
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

42558-54-3
144.17
HNNFDXWDCFCVDM-UHFFFAOYSA-N
2759969
CC(C)C(=O)CC(=O)OC
C7H12O3
MFCD00040499
methyl isobutyrylacetate, methyl 4-methyl-3-oxovalerate, 4-methyl-3-oxovaleric acid methyl ester, iso-butyryl methyl acetate, methyl 4-methyl-3-oxo-pentanoate, methylisobutyrylacetate, acmc-209jq9, ksc237g2d, methyl 4-methyl-3oxopentanoate, methyl4-methyl-3-oxopentanoate
methyl 4-methyl-3-oxopentanoate

Specifications

42558-54-3
1.01
79°C (174°F)
1.4265
1857823
HNNFDXWDCFCVDM-UHFFFAOYSA-N
methyl 4-methyl-3-oxopentanoate
2759969
98%
-75°C
192°C
C7H12O3
MFCD00040499
methyl isobutyrylacetate, methyl 4-methyl-3-oxovalerate, 4-methyl-3-oxovaleric acid methyl ester, iso-butyryl methyl acetate, methyl 4-methyl-3-oxo-pentanoate, methylisobutyrylacetate, acmc-209jq9, ksc237g2d, methyl 4-methyl-3oxopentanoate, methyl4-methyl-3-oxopentanoate
CC(C)C(=O)CC(=O)OC
144.17
144.17
Methyl isobutyrylacetate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

P210-P280-P370+P378q-P501c

H227

EINECSNumber : 418-900-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only