accessibility menu, dialog, popup

UserName

H3CCH3OOOCH3

CAS RN 42558-54-3

IUPAC Name: methyl 4-methyl-3-oxopentanoate
Synonyms: methyl 4-methyl-3-oxopentanoate
Molecular Weight (g/mol): 144.17
Molecular Formula: C7H12O3
InChi Key: HNNFDXWDCFCVDM-UHFFFAOYSA-N
SMILES: COC(=O)CC(=O)C(C)C
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)