missing translation for 'onlineSavingsMsg'
Learn More

Methyl isobutyrylacetate, 97+%

Catalog No. AAL1621622
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1621622 100 g
AAL1621614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1621622 Supplier Thermo Scientific Chemicals Supplier No. L1621622

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 42558-54-3 | C7H12O3 | 144.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 42558-54-3
Molecular Formula C7H12O3
Molecular Weight (g/mol) 144.17
MDL Number MFCD00040499
InChI Key HNNFDXWDCFCVDM-UHFFFAOYSA-N
Synonym methyl isobutyrylacetate, methyl 4-methyl-3-oxovalerate, 4-methyl-3-oxovaleric acid methyl ester, iso-butyryl methyl acetate, methyl 4-methyl-3-oxo-pentanoate, methylisobutyrylacetate, acmc-209jq9, ksc237g2d, methyl 4-methyl-3oxopentanoate, methyl4-methyl-3-oxopentanoate
PubChem CID 2759969
IUPAC Name methyl 4-methyl-3-oxopentanoate
SMILES CC(C)C(=O)CC(=O)OC

Specifications

Melting Point -75°C
Density 1.01
Boiling Point 192°C
Flash Point 79°C (174°F)
Refractive Index 1.4265
Quantity 100 g
Beilstein 1857823
Formula Weight 144.17
Percent Purity 98%
Chemical Name or Material Methyl isobutyrylacetate
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 418-900-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only