missing translation for 'onlineSavingsMsg'
Learn More

Methyl hydrogen suberate, 98%

Catalog No. AAH3032514
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3032514 25 g
AAH3032503 1 g
AAH3032506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH3032514 Supplier Thermo Scientific Chemicals Supplier No. H3032514
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3946-32-5 | C9H15O4 | 187.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3946-32-5
Molecular Formula C9H15O4
Molecular Weight (g/mol) 187.22
MDL Number MFCD00004427
InChI Key KOVPXZDUVJGGFU-UHFFFAOYSA-M
Synonym suberic acid monomethyl ester, monomethyl suberate, methyl hydrogen suberate, subericacidmonomethylester, octanedioic acid monomethyl ester, mono-methyl suberate, acmc-1ckw1, ksc492m6f, 7-methoxycarbonylheptanoic acid, octanedioic acid 1-methyl ester
PubChem CID 554191
IUPAC Name 8-methoxy-8-oxooctanoic acid
SMILES COC(=O)CCCCCCC([O-])=O

Specifications

Melting Point 17°C to 19°C
Density 1.04
Boiling Point 185°C to 186°C (18 mmHg)
Linear Formula CH3O2C(CH2)6CO2H
Quantity 25 g
Formula Weight 188.22
Percent Purity 98%
Chemical Name or Material Methyl hydrogen suberate
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only