missing translation for 'onlineSavingsMsg'
Learn More

Methyl Hexanoate 98.0+%, TCI America™

Supplier:  TCI America H0111500ML

 View more versions of this product

Catalog No. H0111500ML

Only null left

Description

Description

Specifications

Specifications

Methyl Hexanoate
-71°C
150°C
MFCD00009510
3272
NUKZAGXMHTUAFE-UHFFFAOYSA-N
methyl hexanoate
7824
130.19
Liquid
106-70-7
Colorless
C7H14O2
500 mL
methyl caproate, hexanoic acid methyl ester, hexanoic acid, methyl ester, methyl hexoate, methyl capronate, methyl n-hexanoate, methyl hexylate, caproic acid methyl ester, methyl caproate natural, methyl n-hexoate
CCCCCC(=O)OC
130.19
CHEBI:77322
≥98.0% (GC)

Chemical Identifiers

106-70-7
130.19
NUKZAGXMHTUAFE-UHFFFAOYSA-N
7824
methyl hexanoate
C7H14O2
MFCD00009510
methyl caproate, hexanoic acid methyl ester, hexanoic acid, methyl ester, methyl hexoate, methyl capronate, methyl n-hexanoate, methyl hexylate, caproic acid methyl ester, methyl caproate natural, methyl n-hexoate
CHEBI:77322
CCCCCC(=O)OC
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0782

RTECSNumber : MO8401400

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions