missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Hexanoate 98.0+%, TCI America™
$45.57 - $161.38
Chemical Identifiers
| CAS | 106-70-7 |
|---|---|
| Molecular Formula | C7H14O2 |
| Molecular Weight (g/mol) | 130.19 |
| MDL Number | MFCD00009510 |
| InChI Key | NUKZAGXMHTUAFE-UHFFFAOYSA-N |
| Synonym | methyl caproate, hexanoic acid methyl ester, hexanoic acid, methyl ester, methyl hexoate, methyl capronate, methyl n-hexanoate, methyl hexylate, caproic acid methyl ester, methyl caproate natural, methyl n-hexoate |
| PubChem CID | 7824 |
| ChEBI | CHEBI:77322 |
| IUPAC Name | methyl hexanoate |
| SMILES | CCCCCC(=O)OC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
H011125ML
|
N/A | 25 mL |
Each for $45.57
|
|
|||||
|
H0111500ML
|
N/A | 500 mL |
Each for $161.38
|
|
|||||
Chemical Identifiers
| C7H14O2 | |
| MFCD00009510 | |
| methyl caproate, hexanoic acid methyl ester, hexanoic acid, methyl ester, methyl hexoate, methyl capronate, methyl n-hexanoate, methyl hexylate, caproic acid methyl ester, methyl caproate natural, methyl n-hexoate | |
| CHEBI:77322 | |
| CCCCCC(=O)OC |
Specifications
| 106-70-7 | |
| Colorless | |
| C7H14O2 | |
| 3272 | |
| NUKZAGXMHTUAFE-UHFFFAOYSA-N | |
| methyl hexanoate | |
| 7824 | |
| 130.19 | |
| Liquid |
| -71°C | |
| 150°C | |
| MFCD00009510 | |
| methyl caproate, hexanoic acid methyl ester, hexanoic acid, methyl ester, methyl hexoate, methyl capronate, methyl n-hexanoate, methyl hexylate, caproic acid methyl ester, methyl caproate natural, methyl n-hexoate | |
| CCCCCC(=O)OC | |
| 130.19 | |
| CHEBI:77322 | |
| ≥98.0% (GC) | |
| Methyl Hexanoate |
Safety and Handling
EINECSNumber : (2)-0782
RTECSNumber : MO8401400
TSCA : Yes