missing translation for 'onlineSavingsMsg'
Learn More

Methyl hexacosanoate, ≥99% (GC), MilliporeSigma™ Supelco™

Catalog No. 111014738
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014738 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014738 Supplier Merck Emd Millipore Supplier No. 52203100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Methyl hexacosanoate is a compound, which can be isolated from lingwu bituminous coal.

Specifications

CAS 5802-82-4
Shipping Condition Ambient
Physical Form Neat
Linear Formula CH3(CH2)24 COOCH3
MDL Number MFCD00042895
Melting Point 63°C to 66°C
Synonym Hexacosanoic acid methyl ester
Type Analytical Standard
Quantity 100 mg
Formula Weight 410.72
Percent Purity ≥99% (GC)
Grade Analytical Standard
Show More Show Less