missing translation for 'onlineSavingsMsg'
Learn More

Methyl Formate 99.5+%, TCI America™

Catalog No. S02995ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S02995ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S02995ML Supplier TCI America Supplier No. S02995ML

Chemical Identifiers

CAS 107-31-3
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00003291
InChI Key TZIHFWKZFHZASV-UHFFFAOYSA-N
Synonym formic acid, methyl ester, methyl methanoate, methylformiat, formic acid methyl ester, formiate de methyle, methylformiaat, mravencan methylnaty, methylformate, caswell no. 570, hcooch3
PubChem CID 7865
ChEBI CHEBI:77699
IUPAC Name methyl formate
SMILES COC=O

Specifications

Color Colorless
Boiling Point 32°C
Quantity 5 mL
UN Number 1243
Formula Weight 60.05
Percent Purity ≥99.5% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Formate [Standard Material for GC]
EINECSNumber (2)-0677
RTECSNumber LQ8925000
TSCA Yes