missing translation for 'onlineSavingsMsg'
Learn More

Methyl Formate 98.0+%, TCI America™

Catalog No. F0086100ML
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F0086100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F0086100ML Supplier TCI America Supplier No. F0086100ML

Chemical Identifiers

CAS 107-31-3
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00003291
InChI Key TZIHFWKZFHZASV-UHFFFAOYSA-N
Synonym formic acid, methyl ester, methyl methanoate, methylformiat, formic acid methyl ester, formiate de methyle, methylformiaat, mravencan methylnaty, methylformate, caswell no. 570, hcooch3
PubChem CID 7865
ChEBI CHEBI:77699
IUPAC Name methyl formate
SMILES COC=O

Specifications

Color Colorless
Boiling Point 32°C
Quantity 100 mL
UN Number 1243
Formula Weight 60.05
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Formate [for Spectrophotometry]
EINECSNumber (2)-0677
RTECSNumber LQ8925000
TSCA Yes
Recommended Storage Refrigerator