missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Ethyl Ketone, for HPLC, GC and spectrophotometry, >99.5%, Solstice
Specifications
Specifications
| CAS Max % | 100.0000% |
| Density | 0.805 g/cm3 |
| Quantity | 20 L |
| Flash Point | −9°C |
| Packaging | HDPE Nowpak™ |
| UN Number | UN1193 |
| Beilstein | 741880 |
| Boiling Point | 79.64°C |
| Formula Weight | 72.11 |
| Percent Purity | >99.5% |
| Show More |
Safety and Handling
| ShelfLife | 547 days from date of manufacture |
| DOTInformation | Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Ethyl methyl ketone |