missing translation for 'onlineSavingsMsg'
Learn More

Methyl Ethyl Ketone, for HPLC, GC and spectrophotometry, >99.5%, Solstice

Catalog No. 6000117
Change view
Click to view available options
Quantity:
56 L
Packaging:
Returnable Steel Drum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6000117 56 L Returnable Steel Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6000117 Supplier Solstice Supplier No. CS24756

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 78-93-3
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00011648
InChI Key ZWEHNKRNPOVVGH-UHFFFAOYSA-N
Synonym 2-butanone, methyl ethyl ketone, butanone, ethyl methyl ketone, methylethyl ketone, meetco, methyl acetone, methylethylketone, 3-butanone, ethylmethylketon
PubChem CID 6569
ChEBI CHEBI:28398
IUPAC Name butan-2-one
SMILES CCC(C)=O

Specifications

CAS Max % 100.0000%
Density 0.805 g/cm3
Quantity 56 L
Flash Point −9°C
Packaging Returnable Steel Drum
UN Number UN1193
Beilstein 741880
Boiling Point 79.64°C
Formula Weight 72.11
Percent Purity >99.5%
Grade HPLC
Chemical Name or Material Methyl Ethyl Ketone, B&J Brand™, for HPLC, GC and spectrophotometry, >99.5%
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Ethyl methyl ketone