missing translation for 'onlineSavingsMsg'
Learn More

Methyl (E)-4-(Bromomethyl)cinnamate 98.0+%, TCI America™

Catalog No. M28165G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M28165G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M28165G Supplier TCI America Supplier No. M28165G
Only null left

Chemical Identifiers

CAS 88738-86-7
Molecular Formula C11H11BrO2
Molecular Weight (g/mol) 255.111
MDL Number MFCD00460746
InChI Key ZSRCGGBALFGALF-VOTSOKGWSA-N
Synonym (E)-4-(Bromomethyl)cinnamic Acid Methyl Ester
PubChem CID 11658908
IUPAC Name methyl (E)-3-[4-(bromomethyl)phenyl]prop-2-enoate
SMILES COC(=O)C=CC1=CC=C(C=C1)CBr

Specifications

Melting Point 60°C
Color White-Yellow
Boiling Point 163°C
Quantity 5 g
Formula Weight 255.11
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl (E)-4-(Bromomethyl)cinnamate