missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Identifiants chimiques
| CAS | 89-91-8 |
|---|---|
| Molecular Formula | C5H10O4 |
| Molecular Weight (g/mol) | 134.13 |
| MDL Number | MFCD00008484 |
| InChI Key | NZTCVGHPDWAALP-UHFFFAOYSA-N |
| Synonym | methyl dimethoxyacetate, acetic acid, dimethoxy-, methyl ester, dimethoxyacetic acid methyl ester, glyoxylic acid methyl ester dimethyl acetal, acetic acid, 2,2-dimethoxy-, methyl ester, glyoxylic acid, methyl ester, 2-dimethyl acetal, zlchem 519, acmc-209r3k, ksc490c4f, methyl 2,2-dimethoxyethanoate |
| PubChem CID | 66647 |
| IUPAC Name | methyl 2,2-dimethoxyacetate |
| SMILES | COC(C(=O)OC)OC |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantity & Availability | |||||
|
AC414180050
|
N/A |
N/A
|
N/A | ||||||
| Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437. | |||||||||
|
AC414180250
|
N/A |
N/A
|
N/A | ||||||
| Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Identifiants chimiques
| C5H10O4 | |
| MFCD00008484 | |
| methyl dimethoxyacetate, acetic acid, dimethoxy-, methyl ester, dimethoxyacetic acid methyl ester, glyoxylic acid methyl ester dimethyl acetal, acetic acid, 2,2-dimethoxy-, methyl ester, glyoxylic acid, methyl ester, 2-dimethyl acetal, zlchem 519, acmc-209r3k, ksc490c4f, methyl 2,2-dimethoxyethanoate | |
| methyl 2,2-dimethoxyacetate |
Spécifications
| 89-91-8 | |
| 160°C to 163°C | |
| Authentic | |
| Glass Bottle | |
| 1.4040 to 1.406 | |
| MFCD00008484 | |
| methyl dimethoxyacetate, acetic acid, dimethoxy-, methyl ester, dimethoxyacetic acid methyl ester, glyoxylic acid methyl ester dimethyl acetal, acetic acid, 2,2-dimethoxy-, methyl ester, glyoxylic acid, methyl ester, 2-dimethyl acetal, zlchem 519, acmc-209r3k, ksc490c4f, methyl 2,2-dimethoxyethanoate | |
| COC(C(=O)OC)OC | |
| 134.13 | |
| 134.13 | |
| Methyl dimethoxyacetate, 99% |
| 1.0900g/mL | |
| 63°C | |
| 97.5% min. (GC) | |
| C5H10O4 | |
| (CH3O)2CHCOOCH3 | |
| 1.09 | |
| NZTCVGHPDWAALP-UHFFFAOYSA-N | |
| methyl 2,2-dimethoxyacetate | |
| 66647 | |
| 98% |
Sécurité et manipulation
MOISTURE SENSITIVE
EINECSNumber : 201-950-0
TSCA : TSCA
RUO – Research Use Only