missing translation for 'onlineSavingsMsg'
Learn More

Methyl Difluoro(fluorosulfonyl)acetate 97.0+%, TCI America™

Catalog No. F03115G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F03115G 5 g
F031125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F03115G Supplier TCI America Supplier No. F03115G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 680-15-9
Molecular Formula C3H3F3O4S
Molecular Weight (g/mol) 192.108
MDL Number MFCD00144316
InChI Key GQJCAQADCPTHKN-UHFFFAOYSA-N
Synonym methyl 2,2-difluoro-2-fluorosulfonyl acetate, methylfluorosulphonyldifluoroacetate, methyl difluoro fluorosulfonyl acetate, methyl fluorosulfonyldifluoroacetate, methyl fluorosulphonyl difluoro acetate, methyl fluorosulphonyldifluoroacetate, methyl 2-fluorosulfonyl difluoroacetate, fluorosulfonyldifluoroacetic acid methyl ester, 2,2-difluoro-2-fluorosulfonylacetic acid methyl ester, difluoro fluorosulfonyl acetic acid methyl ester
PubChem CID 547667
IUPAC Name methyl 2,2-difluoro-2-fluorosulfonylacetate
SMILES COC(=O)C(F)(F)S(=O)(=O)F

Specifications

Color Yellow
Boiling Point 118°C
Quantity 5 g
UN Number 2920
Formula Weight 192.11
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Difluoro(fluorosulfonyl)acetate