missing translation for 'onlineSavingsMsg'
Learn More

Methyl Dichloroacetate 99.0+%, TCI America™

Catalog No. D0309500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0309500G 500 g
D030925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D0309500G Supplier TCI America Supplier No. D0309500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 116-54-1
Molecular Formula C3H4Cl2O2
Molecular Weight (g/mol) 142.963
MDL Number MFCD00000843
InChI Key HKMLRUAPIDAGIE-UHFFFAOYSA-N
Synonym methyl dichloroacetate, dichloroacetic acid methyl ester, acetic acid, dichloro-, methyl ester, ccris 7740, acetic acid, 2,2-dichloro-, methyl ester, methyldichloroethanoate, acmc-1brpm, dichloroacetic acid methyl, methyl2,2-dichloroacetate, wln: gygvo1
PubChem CID 8315
IUPAC Name methyl 2,2-dichloroacetate
SMILES COC(=O)C(Cl)Cl

Specifications

Melting Point -52°C
Color Yellow
Boiling Point 143°C
Quantity 500 g
UN Number 2299
Formula Weight 142.96
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Dichloroacetate
EINECSNumber (2)-1163
RTECSNumber AG6625000
TSCA Yes