missing translation for 'onlineSavingsMsg'
Learn More

Methyl-beta-D-galactopyranoside, 98%

Catalog No. AC314980050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC314980050 5 g Glass bottle
AC314980010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC314980050 Supplier Thermo Scientific Chemicals Supplier No. 314980050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1824-94-8 | C7H14O6 | 194.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1824-94-8
Molecular Formula C7H14O6
Molecular Weight (g/mol) 194.18
MDL Number MFCD00064357
InChI Key HOVAGTYPODGVJG-VOQCIKJUSA-N
Synonym methyl beta-d-galactopyranoside, methyl beta-d-galactoside, methyl galactoside, methylgalactoside, methyl beta-galactoside, galbetaome, unii-64ryd088rj, beta-d-galactopyranose methyl glycoside, beta-methyl-d-galactoside, methyl-beta-galactose
PubChem CID 94214
ChEBI CHEBI:17540
IUPAC Name (2R,3R,4S,5R,6R)-2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol
SMILES CO[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O

Specifications

Melting Point 175°C to 179°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (ELSD) (HPLC)
Packaging Glass bottle
Quantity 5 g
Specific Rotation Condition −18° (20° (c=1.5,MeOH))
Specific Rotation -18
Solubility Information Solubility in water: soluble in water. Other solubilities: soluble in methanol
Formula Weight 194.18
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Methyl-β-D-galactopyranoside
Show More Show Less
EINECSNumber 217-361-7

RUO – Research Use Only