missing translation for 'onlineSavingsMsg'
Learn More

Methyl Cyclobutanecarboxylate 98.0+%, TCI America™

Catalog No. M28845G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M28845G 5 g
M288425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M28845G Supplier TCI America Supplier No. M28845G

Chemical Identifiers

CAS 765-85-5
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.14
MDL Number MFCD00095142
InChI Key CBTGNLZUIZHUHY-UHFFFAOYSA-N
Synonym cyclobutanecarboxylic acid methyl ester, cyclobutanecarboxylic acid, methyl ester, pubchem23839, methyl cyclobutane carboxylate, cyclobutanecarboxylicacid,methylester, cyclobutanecarboxylicacid, methyl ester, cyclobutane carboxylic acid methyl ester
PubChem CID 136594
IUPAC Name methyl cyclobutanecarboxylate
SMILES COC(=O)C1CCC1

Specifications

Color Colorless
Boiling Point 135°C
Quantity 5 g
UN Number 3272
Formula Weight 114.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Cyclobutanecarboxylate