missing translation for 'onlineSavingsMsg'
Learn More

Methyl cyclobutanecarboxylate, 98%

Catalog No. p-2691775
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC365540050 5 g Glass Bottle
AC365540250 25 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC365540050 Supplier Thermo Scientific Chemicals Supplier No. 365540050

CAS: 765-85-5 | C6H10O2 | 114.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 765-85-5
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.14
MDL Number MFCD00095142
InChI Key CBTGNLZUIZHUHY-UHFFFAOYSA-N
Synonym cyclobutanecarboxylic acid methyl ester, cyclobutanecarboxylic acid, methyl ester, pubchem23839, methyl cyclobutane carboxylate, cyclobutanecarboxylicacid,methylester, cyclobutanecarboxylicacid, methyl ester, cyclobutane carboxylic acid methyl ester
PubChem CID 136594
IUPAC Name methyl cyclobutanecarboxylate
SMILES COC(=O)C1CCC1

Specifications

Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Refractive Index 1.4260 to 1.4300 (20°C, 589nm)
Quantity 5 g
Formula Weight 114.14
Percent Purity 98%
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Methyl cyclobutanecarboxylate, 98%

RUO – Research Use Only